Learn More
Bioworld LPALINEAR POLYACRYLAMIDE)
Catalog No. 50196649
Encompass_Preferred
LPA (LINEAR POLYACRYLAMIDE) FOR DNA/RNA PRECIPITATION: LPA is an inert coprecipitant used to aid recovery of nucleic acids during alcohol precipitations, essential for quantitative recovery of small amounts of nucleic acids in dilute sol...
Catalog No. 50-196-649 Supplier Bioworld Supplier No. 105100002
LPA (LINEAR POLYACRYLAMIDE) FOR DNA/RNA PRECIPITATION: LPA is an inert coprecipitant used to aid recovery of nucleic acids during alcohol precipitations, essential for quantitative recovery of small amounts of nucleic acids in dilute sol...