Learn More
ABclonal Technology LPAR2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A148191000UL
Encompass_Preferred
This gene encodes a member of family I of the G protein-coupled receptors, as well as the EDG family of proteins. This protein functions as a lysophosphatidic acid (LPA) receptor and contributes to Ca2+ mobilization, a critical cellular response to LPA in cells, through association with Gi and Gq proteins. An alternative splice variant has been described but its full length sequence has not been determined.
Catalog No. 50-001-06155