Learn More
Aviva Systems Biology Corp LPAR4 Antibody - C-terminal region (ARP64699_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64699_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LPAR4. It was validated on Western Blot. . Alternate names: GPR23, LPA4, P2RY9, P2Y5-LIKE, P2Y9ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-288-340