Learn More
Adam Equipment LPB 623i - Luna Precision Balances - Capacity: 620g, Readability: 0.001g
Catalog No. NC3964830
Encompass_Preferred
Elevate your laboratory's precision and efficiency with Luna Precision Balances, designed to meet the rigorous demands of labs, manufacturing, and production environments. Luna balances excel in tasks such as density determination, purity analysis, and quality control, offering unmatched durability and performance. The included hook facilitates below-balance weighing, expanding your measurement capabilities. Luna balances are equipped with USB and RS-232 interfaces, ensuring seamless communication with PCs and printers. This connectivity supports data tracking within Good Laboratory Practices (GLP) guidelines.
Catalog No. NC3964830 Supplier Adam Equipment Supplier No. LPB 623I
Elevate your laboratory's precision and efficiency with Luna Precision Balances, designed to meet the rigorous demands of labs, manufacturing, and production environments. Luna balances excel in tasks such as density determination, purity analysis, and quality control, offering unmatched durability and performance. The included hook facilitates below-balance weighing, expanding your measurement capabilities. Luna balances are equipped with USB and RS-232 interfaces, ensuring seamless communication with PCs and printers. This connectivity supports data tracking within Good Laboratory Practices (GLP) guidelines.