Learn More
Adam Equipment Adam Equipment LPB 823e Luna Precision Balance 820g Capacity x 0.001g Readability
Catalog No. NC2021387
Encompass_Preferred
Adam Equipment's Luna precision balances are suitable for most lab weighing tasks. Luna’s display features a stylish look, with easy-to-read 24mm digits in blue on a black background. Ideal for international use, Luna provides multilingual text and display capabilities. USB and RS-232 interfaces facilitate data gathering and enable smooth communication with printers and computers. The grade 304 stainless steel pan and sturdy ABS plastic construction are easy to keep clean, giving Luna the rugged durability needed in a lab balance.
Catalog No. NC2021387 Supplier Adam Equipment Supplier No. LPB 823E
Adam Equipment's Luna precision balances are suitable for most lab weighing tasks. Luna’s display features a stylish look, with easy-to-read 24mm digits in blue on a black background. Ideal for international use, Luna provides multilingual text and display capabilities. USB and RS-232 interfaces facilitate data gathering and enable smooth communication with printers and computers. The grade 304 stainless steel pan and sturdy ABS plastic construction are easy to keep clean, giving Luna the rugged durability needed in a lab balance.