Learn More
Cayman Chemical LpBromotetramIsoleoxalate 10mg
SDP
Catalog No. 502967341
Encompass_Preferred
A cell-permeable inhibitor of all four human AP isoenzymes (Kis =18 and 56 µM for PLAP and NSAP, respectively); also used as a tyrosine phosphatase inhibitor
Catalog No. 50-296-7341 Supplier Cayman Chemical Supplier No. 1871310 MG
A cell-permeable inhibitor of all four human AP isoenzymes (Kis =18 and 56 µM for PLAP and NSAP, respectively); also used as a tyrosine phosphatase inhibitor