Learn More
MSC LPD Cut-Resistant Gloves, Cut A5, ESD, Size Large, 1 Pair
Catalog No. 5000677005
Encompass
These cut-resistant gloves are made from Ultra High Molecular Weight Polyethylene (UHMW-PE) with a polyurethane coating on the palm and fingers. They provide ANSI/ISEA Cut Resistance Level A5 and are static dissipative with ESD features, making them suitable for environments where electrostatic discharge is a concern. The gloves are size Large and come as a pair.
  • Made from ultra high molecular weight polyethylene (UHMW-PE)
  • Polyurethane coating on palm and fingers
  • ANSI/ISEA Cut Resistance Level A5
  • ANSI/ISEA Puncture Resistance Level 0
  • ANSI/ISEA Abrasion Resistance Level 0
  • Static dissipative with ESD features
  • Glove size: Large (numeric size 9)
  • Knit style with dipped coating
  • Unisex design
  • Country of origin: Malaysia
Catalog No. 50-006-77005 Supplier MSC Supplier No. 20439576
These cut-resistant gloves are made from Ultra High Molecular Weight Polyethylene (UHMW-PE) with a polyurethane coating on the palm and fingers. They provide ANSI/ISEA Cut Resistance Level A5 and are static dissipative with ESD features, making them suitable for environments where electrostatic discharge is a concern. The gloves are size Large and come as a pair.
  • Made from ultra high molecular weight polyethylene (UHMW-PE)
  • Polyurethane coating on palm and fingers
  • ANSI/ISEA Cut Resistance Level A5
  • ANSI/ISEA Puncture Resistance Level 0
  • ANSI/ISEA Abrasion Resistance Level 0
  • Static dissipative with ESD features
  • Glove size: Large (numeric size 9)
  • Knit style with dipped coating
  • Unisex design
  • Country of origin: Malaysia