Learn More
Cayman Chemical Lipid HTO12, Purity: ≥98%, CAS: 3047222-45-4, 5 mg
SDP
Catalog No. 502944294
Encompass_Preferred
An ionizable cationic lipid (pKa = 6.432); has been used in combination with other lipids in the formation of LNPs for the delivery of mRNA in vitro and in vivo; LNPs containing lipid HTO12 and encapsulating adenine base editor mRNA targeting Pcsk9 reduce hepatic Pcsk9 levels in mice at a reduced ionizable lipid/total lipids-to-mRNA ratio than LNPs containing SM-102
Catalog No. 50-294-4294 Supplier Cayman Chemical Supplier No. 412025 MG
An ionizable cationic lipid (pKa = 6.432); has been used in combination with other lipids in the formation of LNPs for the delivery of mRNA in vitro and in vivo; LNPs containing lipid HTO12 and encapsulating adenine base editor mRNA targeting Pcsk9 reduce hepatic Pcsk9 levels in mice at a reduced ionizable lipid/total lipids-to-mRNA ratio than LNPs containing SM-102