Learn More
MilliporeSigma™ Supelco™ LpDNPH H Series Cartridges H10
Catalog No. 111009671 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Volume (Metric):
3 mL
Catalog No. Volume (Metric)
11-100-9671 3 mL
1 options
Catalog No. 11-100-9671 Supplier MilliporeSigma™ Supelco™ Supplier No. 505315
Add to Cart
Add to Cart
Used for sampling carbonyls
LpDNPH H Series Cartridges (H10) are air sampling devices used for sampling carbonyls via trapping them on a high purity silica adsorbent coated with 2,4-dinitrophenylhydrazine (2,4-DNPH), where they are converted to the hydrazone derivatives. The derivatives are eluted from the cartridge in acetonitrile and analyzed by HPLC. The built-in reservoir eliminates the need to attach a syringe to the cartridge for sample extraction/elution. Reusable adapters are available to connect the cartridge to the sampling pump.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10/Pk.
Volume (Metric) 3 mL