Learn More
MilliporeSigma™ Supelco™ LpDNPH Rezorian Cartridge, Bed wt. 350 mg
Catalog No. 111007703 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10/Pk.
50/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-7703 50/Pk.
11-101-0235 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-7703 Supplier MilliporeSigma™ Supelco™ Supplier No. 54075U
Only null left
Add to Cart
Add to Cart
Solvent-compatible due to its polypropylene components and polyethylene frits
LpDNPH Rezorian™ Cartridge are solvent-compatible due to its polypropylene components and polyethylene frits. It can be commonly used, either individually or connected in series. It is employed in-line with peristaltic pumps and low pressure HPLC systems, or with a syringe and a single sample processor.
TRUSTED_SUSTAINABILITY
Specifications
Description Bed wt. 350 mg
Quantity 50/Pk.
Volume (Metric) 3 mL