Learn More
MilliporeSigma™ Supelco™ LpDNPH S10 Cartridge, Bed wt. 350 mg
Catalog No. 111016363 Shop All MilliporeSigma Supelco Products
Click to view available options
Type:
S10 Cartridge
SPE Cartridge
Quantity:
10/Pk.
50/Pk.
Packaging:
1/Zip Bag
Bulk Packed
Easy to use in the field and laboratory
The design of the S10 cartridge makes it easy to use in the field and laboratory. Reusable adaptors are available for connecting the cartridge to the sampling pump. The built in reservoir eliminates the need to attach a syringe for sample extraction/elution.
TRUSTED_SUSTAINABILITY
Specifications
Basis Weight 350 mg
Certifications/Compliance ASTM D5197/EPA 100,IP-6 A,TO-11 A/NIOSH 2016
Packaging Bulk Packed
Quantity 50/Pk.
Type S10 Cartridge
Volume (Metric) 3 mL
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.