Learn More
MilliporeSigma™ Supelco™ LpDNPH S10 Cartridge Starter Kit, Bed wt. 350 mg
Catalog No. 111010059 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Volume (Metric):
3 mL
Catalog No. Volume (Metric)
11-101-0059 3 mL
1 options
Catalog No. 11-101-0059 Supplier MilliporeSigma™ Supelco™ Supplier No. 21024U
Add to Cart
Add to Cart
Easy to use in the field and laboratory
The design of the S10 cartridge makes it easy to use in the field and laboratory. Reusable adaptors are available for connecting the cartridge to the sampling pump. The built in reservoir eliminates the need to attach a syringe for sample extraction/elution.
TRUSTED_SUSTAINABILITY
Specifications
Description Bed wt. 350 mg
Quantity 10/Pk.
Volume (Metric) 3 mL