Learn More
MilliporeSigma™ Supelco™ LpDNPH S10L Cartridge, Bed wt. 350 mg
Catalog No. 111020811 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Volume (Metric):
1 mL
Catalog No. Volume (Metric)
11-102-0811 1 mL
1 options
Catalog No. 11-102-0811 Supplier MilliporeSigma™ Supelco™ Supplier No. 505358
Add to Cart
Add to Cart
Meets EPA TO-11A requirements.
The S10L cartridges features a reversible design and is for analysts who prefer shorter dimensions and does not require an adaptor for sampling. The cartridge is eluted by connecting to a syringe barrel (empty SPE cartridge) that acts as a reservoir for gravity-fed elution solvent. Meets EPA TO-11A requirements.
TRUSTED_SUSTAINABILITY
Specifications
Description Bed wt. 350 mg
Quantity 10/Pk.
Volume (Metric) 1 mL