Learn More
Aviva Systems Biology Corp LPGAT1 Antibody - C-terminal region (ARP62465_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62465_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LPGAT1. It was validated on Western Blot. . Alternate names: FAM34A, FAM34A1, NET8ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-287-690