Learn More
Cayman Chemical LPhenylephrInhydroCLOrId 100mg
SDP
Catalog No. 502966226
Encompass_Preferred
An adrenergic α1A receptor agonist (Ki = 1.4 µM) that demonstrates selectivity against the α1B and α1C receptor subtypes (Kis = 23.9 and 47.8 µM, respectively); frequently used to precontract smooth muscle in preparations designed to study the properties of various vasodilator agents
Catalog No. 50-296-6226 Supplier Cayman Chemical Supplier No. 18619100 MG
An adrenergic α1A receptor agonist (Ki = 1.4 µM) that demonstrates selectivity against the α1B and α1C receptor subtypes (Kis = 23.9 and 47.8 µM, respectively); frequently used to precontract smooth muscle in preparations designed to study the properties of various vasodilator agents