Learn More
Aviva Systems Biology Corp LPIN1 Antibody - N-terminal region (ARP53826_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53826_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LPIN1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LPIN1. Alternate names: DKFZp781P1796, KIAA0188, PAP1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Yeast, Zebrafish
Catalog No. 50-014-53731