Learn More
Aviva Systems Biology Corp LPIN2 Antibody - C-terminal region (ARP61571_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61571_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LPIN2. It was validated on Western Blot. . Alternate names: KIAA0249ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-410