Learn More
Echelon Biosciences Research Labs LPLA2 ENZYME 10ug
SDP
Catalog No. 5082225
Encompass
Recombinant full-length human lysosomal phospholipase A2 (LPLA2) protein with C-terminal 6X-HIS tag was expressed in HEK 293 cells and purified using Ni-IMAC chromatography. The 6X-His tag was then removed by TEV protease.hLPLA2 is a lysosomal enzyme that has both calcium-independent phospholipase A2 and transacylase activities. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-822-25 Supplier Echelon Biosciences Research Labs Supplier No. E700010UG
Recombinant full-length human lysosomal phospholipase A2 (LPLA2) protein with C-terminal 6X-HIS tag was expressed in HEK 293 cells and purified using Ni-IMAC chromatography. The 6X-His tag was then removed by TEV protease.hLPLA2 is a lysosomal enzyme that has both calcium-independent phospholipase A2 and transacylase activities. This item is non returnable