Learn More
Sigma Aldrich Fine Chemicals Biosciences LpodsqR StyreneMalec A100MG
Catalog No. 502904312
Encompass
Lipodisq(R) polymers contain styrene and maleic acid monomer units. This product is the pre-hydrolyzed form of the SMA polymer with the styrene and maleic acid monomers. These polymer-lipid complexes are mainly generated by detergent-free methods.
Catalog No. 50-290-4312 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L9170100MG
Lipodisq(R) polymers contain styrene and maleic acid monomer units. This product is the pre-hydrolyzed form of the SMA polymer with the styrene and maleic acid monomers. These polymer-lipid complexes are mainly generated by detergent-free methods.