Learn More
Aviva Systems Biology Corp PLPPR2 Antibody - middle region (ARP49743_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49743_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LPPR2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LPPR2. Alternate names: DKFZp761E1121, FLJ13055, PRG-4, PRG4ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53599