Learn More
Invivogen LPS-EB Ultrapure, 5mg Lipopolysaccharide from E. coli
SDP
Supplier:  Invivogen TLRL3PELPS
Encompass
5mg lipopolysaccharide from E. coli 0111:B4 (LPS-EB). 1.5mL endotoxin-free water. Shipped at room temperature and should be stored at 4 degrees Celsius. Product is stable for 1 year at -20 degrees Celsius. Unit size: 5x10e6 EU.
Catalog No. NC0202558