Learn More
Boca Scientific Inc LPS Extraction Kit, 100 Extractions
Catalog No. NC9753815
Encompass
The LPS Extraction Kit is designed to separate and purify Lipopolysaccharides (LPS) present in the outer wall of gram-negative bacteria. This kit can extract LPS from even a small volume of bacterial cells and gives reproducibly high yields of LPS. The protocol takes less than 60 minutes and includes only three steps: lysis of bacterial cells, purification of LPS among the released cell components, and washing to remove salts. This kit provides 100 extractions.
Catalog No. NC9753815 Supplier Boca Scientific Inc Supplier No. 17141
Add to Cart
Add to Cart
The LPS Extraction Kit is designed to separate and purify Lipopolysaccharides (LPS) present in the outer wall of gram-negative bacteria. This kit can extract LPS from even a small volume of bacterial cells and gives reproducibly high yields of LPS. The protocol takes less than 60 minutes and includes only three steps: lysis of bacterial cells, purification of LPS among the released cell components, and washing to remove salts. This kit provides 100 extractions.