Learn More
AdipoGen LPS from E. coli O111:B4 (S-form) TLRpure Sterile Solution, 1 mg
SDP
Supplier:  AdipoGen IAX100012M001
Encompass_Preferred
≥99.9%, No detectable DNA, RNA and protein traces. Formulation: Liquid. Colourless clear aqueous solution. Concentration: 1mg/ml stabilised in sterile, double-distilled water (ddWater), without any additives. LPS are amphipathic molecules whose hydrophobicity decreases with increasing length of the sugar part. Based upon these differences, S- and R-form LPS show marked differences in the kinetics of their blood clearance and cellular uptake as well as in the ability to induce oxidative burst in human granulocytes and to activate the host complement system.
Catalog No. 50-596-699