Learn More
Enzo Life Sciences LPS from E. coli, Serotype O55:B5 (TLRGRADE) (Ready-to-Use), (1ml)
SDP
Catalog No. NC1198683
Encompass_Preferred
Specific TLR4 activator. Alternative Name: Lipopolysaccharide from E. coli, Serotype O55:B5 S-form. Purity: Absence of detectable protein or DNA contaminants with agonistic TLR activity. Use/Stability: Stable for at least 1 year after receipt when stored at +4°C. Long Term Storage: +4°C
Catalog No. NC1198683 Supplier Enzo Life Sciences Supplier No. ALX581013L001
Specific TLR4 activator. Alternative Name: Lipopolysaccharide from E. coli, Serotype O55:B5 S-form. Purity: Absence of detectable protein or DNA contaminants with agonistic TLR activity. Use/Stability: Stable for at least 1 year after receipt when stored at +4°C. Long Term Storage: +4°C