Learn More
AdipoGen LPS from S. abortus equi (S-form) Biotin TLRpure Sterile Solution, 500 ug
SDP
Supplier:  AdipoGen IAX100009BC500
Encompass_Preferred
≥99.9%, No detectable DNA, RNA and protein traces. Formulation: Liquid. Colourless clear aqueous solution. Concentration: 1mg/ml (0.5 mg/ml for 250mg size) stabilised in sterile, double-distilled water (ddWater), without any additives. LPS are amphipathic molecules whose hydrophobicity decreases with increasing length of the sugar part. Based upon these differences, S- and R-form LPS show marked differences in the kinetics of their blood clearance and cellular uptake as well as in the ability to induce oxidative burst in human granulocytes and to activate the host complement system.
Catalog No. 50-117-6313