Learn More
Enzo Life Sciences LPS from Salmonella abortus equi S-form (TLRGRADE™) (Ready-to-Use) (Biotin) (500µl)
SDP
Catalog No. 502009576
Encompass_Preferred
Useful reagent for evaluating LPS binding and cell activation in white blood cells. Alternative name: Lipopolysaccharide from Salmonella abortus equi S-form. Formulation: Liquid. Sterile, ready-to-use solution in pyrogen-free double-distilled water. Purity: Absence of dectectable protein or DNA contaminants with agonistic TLR activity. Long Term Storage: +4°C.
Catalog No. 50-200-9576 Supplier Enzo Life Sciences Supplier No. ALX581150R500
Useful reagent for evaluating LPS binding and cell activation in white blood cells. Alternative name: Lipopolysaccharide from Salmonella abortus equi S-form. Formulation: Liquid. Sterile, ready-to-use solution in pyrogen-free double-distilled water. Purity: Absence of dectectable protein or DNA contaminants with agonistic TLR activity. Long Term Storage: +4°C.