Learn More
Sigma Aldrich Fine Chemicals Biosciences Lipopolysaccharides from Salmonella enterica serotype typhimurium, MFCD00164404, 1 mg
Catalog No. NC1086121
Encompass
BioXtra, suitable for cell culture, γ-irradiated, Synonym: LPS. Lipopolysaccharides from Salmonella enterica serotype typhimurium has been used: in mice to elicit the secretion of pro-inflammatory cytokines, to induce cytokine production in monocytes, to elicit the activation of NF-κB pathway in mouse dendritic cells, to induce inflammatory response in human colon adenocarcinoma Caco-2 cells, and for stimulation of the catfish peripheral blood leukocytes (PBLs) to study estrogen receptor mRNA expression.
Catalog No. NC1086121 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L61431MG
BioXtra, suitable for cell culture, γ-irradiated, Synonym: LPS. Lipopolysaccharides from Salmonella enterica serotype typhimurium has been used: in mice to elicit the secretion of pro-inflammatory cytokines, to induce cytokine production in monocytes, to elicit the activation of NF-κB pathway in mouse dendritic cells, to induce inflammatory response in human colon adenocarcinoma Caco-2 cells, and for stimulation of the catfish peripheral blood leukocytes (PBLs) to study estrogen receptor mRNA expression.