Learn More
Enzo Life Sciences LPS from Salmonella minnesota R595 (Re) (TLRGRADE) (Ready-to-Use), (2ml)
SDP
Supplier:  Enzo Life Sciences ALX581008L002
Encompass_Preferred
TLR4 activator. Alternative Name: Lipopolysaccharide from Salmonella minnesota R595 (Re). Purity: Absence of detectable protein or DNA contaminants with agonistic TLR activity. Use/Stability: Stable for at least 1 year after receipt when stored at +4°C. Long Term Storage: +4°C
Catalog No. NC9344660