Learn More
Enzo Life Sciences LPS from Salmonella minnesota R5 (Rc) (TLRGRADE™) (Ready-to-Use) (2ml)
SDP
Supplier:  Enzo Life Sciences ALX581017L002
Encompass_Preferred
Alternative name: Lipopolysaccharide from Salmonella minnesota R5 (Rc). Formulation: Liquid. Sterile, ready-to-use solution in pyrogen-free double distilled water. Purity: Absence of detectable protein or DNA contaminants with agonistic TLR activity. Long Term Storage: +4°C.
Catalog No. 50-200-9572