Learn More
Enzo Life Sciences LPS from Salmonella typhimurium S-form (TLRGRADE™) (Ready-to-Use) (1ml)
SDP
Catalog No. 502009568
Encompass_Preferred
Alternative name: Lipopolysaccharide from Salmonella typhimurium S-form. Formulation: Liquid. Sterile, ready-to-use solution in pyrogen-free double distilled water. Purity: Absence of detectable protein or DNA contaminants with agonistic TLR activity. Long Term Storage: +4°C.
Catalog No. 50-200-9568 Supplier Enzo Life Sciences Supplier No. ALX581011L001
Alternative name: Lipopolysaccharide from Salmonella typhimurium S-form. Formulation: Liquid. Sterile, ready-to-use solution in pyrogen-free double distilled water. Purity: Absence of detectable protein or DNA contaminants with agonistic TLR activity. Long Term Storage: +4°C.