Learn More
MSC LPS Cleaner Degreaser, Concentrated, Unscented, NSF A1, 28 oz
Catalog No. 502867888
Encompass
LPS T-91 Cleaner & Degreaser is a concentrated, pH neutral degreaser formulated to effectively remove grease, oil, and other contaminants from a wide range of surfaces. This unscented, ready-to-use liquid is suitable for various industrial and marine applications, meeting NSF A1 standards for safety and performance.
  • Concentrated liquid formulation
  • Effectively removes oil and grease
  • Unscented for versatile use
  • Suitable for use in basin style parts washers, bilges, and dip tanks
  • Meets NSF A1 standards
Catalog No. 50-286-7888 Supplier MSC Supplier No. 77679678
LPS T-91 Cleaner & Degreaser is a concentrated, pH neutral degreaser formulated to effectively remove grease, oil, and other contaminants from a wide range of surfaces. This unscented, ready-to-use liquid is suitable for various industrial and marine applications, meeting NSF A1 standards for safety and performance.
  • Concentrated liquid formulation
  • Effectively removes oil and grease
  • Unscented for versatile use
  • Suitable for use in basin style parts washers, bilges, and dip tanks
  • Meets NSF A1 standards