Learn More
Sarstedt Inc LPSCMT,1.5ML,CBSGWB,ASM/PK1000
Supplier:  Sarstedt Inc 72.703.416
Encompass_Preferred
1.5ml Loop Screw Cap Micro Tube Conical Bottom Skirted Graduations and Writing Block Assembled PCR Performance Tested
Catalog No. NC0069871