Learn More
Medchemexpress LLC LpxC-IN-10 | 2413574-64-6 | 95.1% | 509.60 | 50 MG
SDP
Catalog No. 5000361429
Encompass_Preferred
LpxC-IN-10 (Compound A) is a highly selective inhibitor of LpxC. It demonstrates MIC values of 0.5 μg/mL against both E. coli and K. pneumoniae. This compound is suitable for research into bacterial infections and functions as a click chemistry reagent, featuring an Alkyne group that allows it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • High selectivity inhibitor of LpxC.
  • Exhibits MIC values of 0.5 μg/mL against E. coli and K. pneumoniae.
  • Useful for bacterial infection research.
  • Click chemistry reagent containing an Alkyne group.
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Azide groups.
Catalog No. 50-003-61429 Supplier Medchemexpress LLC Supplier No. HY14723750MG
LpxC-IN-10 (Compound A) is a highly selective inhibitor of LpxC. It demonstrates MIC values of 0.5 μg/mL against both E. coli and K. pneumoniae. This compound is suitable for research into bacterial infections and functions as a click chemistry reagent, featuring an Alkyne group that allows it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • High selectivity inhibitor of LpxC.
  • Exhibits MIC values of 0.5 μg/mL against E. coli and K. pneumoniae.
  • Useful for bacterial infection research.
  • Click chemistry reagent containing an Alkyne group.
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Azide groups.