Learn More
TARGETMOL CHEMICALS INC LPXH-IN-AZ1 5MG
SDP
Catalog No. 502577440
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. LpxH-IN-AZ1 is a potent inhibitor of the UDP-23-diacylglucosamine pyrophosphate hydrolase LpxH and sulfonylpiperazine compounds.LpxH-IN-AZ1 exhibits antimicrobial activity and inhibits Klebsiella pneumoniae with an IC50 of 0.36 uM. purity: 99%
Catalog No. 50-257-7440 Supplier TARGETMOL CHEMICALS INC Supplier No. T118765MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. LpxH-IN-AZ1 is a potent inhibitor of the UDP-23-diacylglucosamine pyrophosphate hydrolase LpxH and sulfonylpiperazine compounds.LpxH-IN-AZ1 exhibits antimicrobial activity and inhibits Klebsiella pneumoniae with an IC50 of 0.36 uM. purity: 99%