Learn More
Uline Liquid Wrench White Lithium Lubricant | White | 1 EA
Catalog No. NC2588994
Encompass_Preferred
Outdoor lubrication that wont freeze melt or wash off, Reduces friction for metal-to-metal contact like hitches hinges and rails, Operates in sub-zero temps down to -40F, Rust inhibitor
Catalog No. NC2588994 Supplier Uline Supplier No. S25326
Outdoor lubrication that wont freeze melt or wash off, Reduces friction for metal-to-metal contact like hitches hinges and rails, Operates in sub-zero temps down to -40F, Rust inhibitor