Learn More
Medchemexpress LLC LQZ-7I | 195822-23-2 | 99.78% | 348.35 | 25 MG
SDP
Catalog No. 5000352327
Encompass_Preferred
LQZ-7I is a survivin-targeting inhibitor that inhibits survivin dimerization. It orally and effectively inhibits xenograft tumor growth and induces survivin loss in tumors.
  • Inhibits survivin dimerization
  • Effectively inhibits xenograft tumor growth
  • Induces survivin loss in tumors
  • Suitable for research use only
Catalog No. 50-003-52327 Supplier Medchemexpress LLC Supplier No. 25MGHY136538
LQZ-7I is a survivin-targeting inhibitor that inhibits survivin dimerization. It orally and effectively inhibits xenograft tumor growth and induces survivin loss in tumors.
  • Inhibits survivin dimerization
  • Effectively inhibits xenograft tumor growth
  • Induces survivin loss in tumors
  • Suitable for research use only