Learn More
Medchemexpress LLC LQZ-7I | 195822-23-2 | 99.8% | 348.35 | 1 ML
SDP
Catalog No. 5000348316
Encompass_Preferred
LQZ-7I is a survivin-targeting inhibitor that inhibits survivin dimerization. It effectively inhibits xenograft tumor growth orally and induces survivin loss in tumors. This compound is for research use only.
  • Inhibits survivin dimerization.
  • Suppresses tumor growth in male NSG mice without notable adverse effects.
  • Exhibits improved cytotoxicity with IC50s of 3.1 μM against C4-2 cells and 4.8 μM against PC-3 cells.
  • Selectively reduces survivin expression without affecting XIAP, cIAP1, and cIAP2.
Catalog No. 50-003-48316 Supplier Medchemexpress LLC Supplier No. HY13653810MM/1ML
LQZ-7I is a survivin-targeting inhibitor that inhibits survivin dimerization. It effectively inhibits xenograft tumor growth orally and induces survivin loss in tumors. This compound is for research use only.
  • Inhibits survivin dimerization.
  • Suppresses tumor growth in male NSG mice without notable adverse effects.
  • Exhibits improved cytotoxicity with IC50s of 3.1 μM against C4-2 cells and 4.8 μM against PC-3 cells.
  • Selectively reduces survivin expression without affecting XIAP, cIAP1, and cIAP2.