Learn More
Medchemexpress LLC LQZ-7I | 195822-23-2 | 99.8% | 348.35 | 100 MG
SDP
Catalog No. 5000351906
Encompass_Preferred
LQZ-7I is a survivin-targeting inhibitor that inhibits survivin dimerization. It orally effectively inhibits xenograft tumor growth and induces survivin loss in tumors.
  • Inhibits survivin dimerization
  • Effectively inhibits xenograft tumor growth
  • Induces survivin loss in tumors
  • Improved cytotoxicity against C4-2 and PC-3 cells
  • Selectively reduces survivin expression
  • Suppresses tumor growth in animal models without adverse effects
Catalog No. 50-003-51906 Supplier Medchemexpress LLC Supplier No. HY136538100MG
LQZ-7I is a survivin-targeting inhibitor that inhibits survivin dimerization. It orally effectively inhibits xenograft tumor growth and induces survivin loss in tumors.
  • Inhibits survivin dimerization
  • Effectively inhibits xenograft tumor growth
  • Induces survivin loss in tumors
  • Improved cytotoxicity against C4-2 and PC-3 cells
  • Selectively reduces survivin expression
  • Suppresses tumor growth in animal models without adverse effects