Learn More
Medchemexpress LLC Lqz-7i | 195822-23-2 | 99.8% | 348.35 g/mol | C20H14F2N4 | 5 MG
SDP
Catalog No. 5000222617
Encompass_Preferred
LQZ-7I is a small-molecule survivin-targeting inhibitor used in research to disrupt survivin dimerization, promote survivin degradation, and reduce tumor growth in xenograft models. It is used in cellular assays and in vivo studies to investigate survivin-related apoptosis pathways.
  • Inhibits survivin dimerization
  • Promotes survivin degradation and induces apoptosis
  • Orally active in xenograft tumor models
  • Suitable for cellular and in vivo research applications
  • Reported purity approximately 99.8%
  • Molecular formula C20H14F2N4; molecular weight 348.35 g/mol
Catalog No. 50-002-22617 Supplier Medchemexpress LLC Supplier No. HY1365385MG
LQZ-7I is a small-molecule survivin-targeting inhibitor used in research to disrupt survivin dimerization, promote survivin degradation, and reduce tumor growth in xenograft models. It is used in cellular assays and in vivo studies to investigate survivin-related apoptosis pathways.
  • Inhibits survivin dimerization
  • Promotes survivin degradation and induces apoptosis
  • Orally active in xenograft tumor models
  • Suitable for cellular and in vivo research applications
  • Reported purity approximately 99.8%
  • Molecular formula C20H14F2N4; molecular weight 348.35 g/mol