Learn More
Medchemexpress LLC Lqz-7i | 195822-23-2 | 99.8% | C20H14F2N4 | 10MG
SDP
Catalog No. 5000261510
Encompass_Preferred
LQZ-7I is a small-molecule survivin-targeting inhibitor that disrupts survivin dimerization, induces proteasome-mediated survivin degradation, and has demonstrated oral antitumor activity in xenograft models. It is supplied as a high-purity solid with documented solubility and storage conditions for use in cellular and in vivo oncology research.
  • Inhibits survivin dimerization and promotes survivin degradation.
  • Demonstrates oral efficacy in xenograft tumor models.
  • High reported purity suitable for research applications.
  • Soluble in DMSO; in vivo formulation protocols available.
  • Available in multiple small pack sizes for preclinical studies.
Catalog No. 50-002-61510 Supplier Medchemexpress LLC Supplier No. HY13653810MG
LQZ-7I is a small-molecule survivin-targeting inhibitor that disrupts survivin dimerization, induces proteasome-mediated survivin degradation, and has demonstrated oral antitumor activity in xenograft models. It is supplied as a high-purity solid with documented solubility and storage conditions for use in cellular and in vivo oncology research.
  • Inhibits survivin dimerization and promotes survivin degradation.
  • Demonstrates oral efficacy in xenograft tumor models.
  • High reported purity suitable for research applications.
  • Soluble in DMSO; in vivo formulation protocols available.
  • Available in multiple small pack sizes for preclinical studies.