Learn More
Selleck Chemical LLC LQZ-7I-E0108-25MG
SDP
Catalog No. 503134708
Encompass_Preferred
LQZ-7I is a novel selectively inhibitor of survivin dimerization with IC50s of 4 8 M and 3 1 M in prostate cancer cell lines PC-3 and C4-2 respectively LQZ-7I orally effectively inhibits xenograft tumor growth and induces survivin loss in tumors
Catalog No. 50-313-4708 Supplier Selleck Chemical LLC Supplier No. E010825MG
LQZ-7I is a novel selectively inhibitor of survivin dimerization with IC50s of 4 8 M and 3 1 M in prostate cancer cell lines PC-3 and C4-2 respectively LQZ-7I orally effectively inhibits xenograft tumor growth and induces survivin loss in tumors