Learn More
Sino Biological LR Cloning Enzyme Kit 100 Tests
Catalog No. 5000696248
Encompass_Preferred
The LR Cloning Enzyme Kit promotes in vitro recombination between attL- and attR-flanked regions on entry clones and destination vectors to generate attB-containing expression clones.
Catalog No. 50-006-96248 Supplier Sino Biological Supplier No. SBLR01100
The LR Cloning Enzyme Kit promotes in vitro recombination between attL- and attR-flanked regions on entry clones and destination vectors to generate attB-containing expression clones.