Learn More
Electron Microscopy Sciences LR Gold Embedding Kit
SDP
Catalog No. 50980417
Encompass_Preferred
The London Resin Company's acrylic "LR Gold" offers the histochemist and immunocytochemist the advantage of working with unfixed tissues at low temperatures, thus allowing many fixation-sensitive procedures to be carried out in 1-4 microns resin sections at -25 degree C using visible light. It is non-toxic and has a low viscosity. Consists of: 500ml LR Gold Resin, 100g Polyvinylpyrolidone; 50g Benz.
Catalog No. 50-980-417 Supplier Electron Microscopy Sciences Supplier No. 14370
The London Resin Company's acrylic "LR Gold" offers the histochemist and immunocytochemist the advantage of working with unfixed tissues at low temperatures, thus allowing many fixation-sensitive procedures to be carried out in 1-4 microns resin sections at -25 degree C using visible light. It is non-toxic and has a low viscosity. Consists of: 500ml LR Gold Resin, 100g Polyvinylpyrolidone; 50g Benz.