Learn More
Hach Company Hardness Quality Control Standard, Low Range, 500 mL
Supplier:  Hach Company 2833449
Encompass
Ready to use - no dilutions required. Mixed standard includes: Total Hardness: 100 mg/L as CaCO3. Calcium Hardness: 50 mg/L as CaCO3.
Catalog No. 50-199-6739