Learn More
Electron Microscopy Sciences LR White Resin Hard - Grade Kit
SDP
Catalog No. 50980422
Encompass_Preferred
A very low viscosity (8cps), non-toxic resin, suitable for both Light and Electron Microscopy. LR White is a polar monomer polyhydroxylated acromatic acrylic resin. It can be cured by heat or by UV light. Sections of polymerized LR White resin are hydrophilic. This character allows immuno-cytochemistry reagents to easily penetrate into the sections and etching of the sections is not necessary (etching sections will effect delicate tissue antigens). LR White sections also show minimal non-specific staining. Its low viscosity makes it ideal for infiltrating decalcified bone and teeth as well as plant tissues. It is available in medium and hard grade. Kit consists of: 500 ml LR White Resin, 10 ml UV accelerator.
Catalog No. 50-980-422 Supplier Electron Microscopy Sciences Supplier No. 14382
A very low viscosity (8cps), non-toxic resin, suitable for both Light and Electron Microscopy. LR White is a polar monomer polyhydroxylated acromatic acrylic resin. It can be cured by heat or by UV light. Sections of polymerized LR White resin are hydrophilic. This character allows immuno-cytochemistry reagents to easily penetrate into the sections and etching of the sections is not necessary (etching sections will effect delicate tissue antigens). LR White sections also show minimal non-specific staining. Its low viscosity makes it ideal for infiltrating decalcified bone and teeth as well as plant tissues. It is available in medium and hard grade. Kit consists of: 500 ml LR White Resin, 10 ml UV accelerator.