Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC LR3 IGF-I/IGF-1 Hum 10ug
SDP

Catalog No. 5000184132
Encompass_Preferred

Recombinant LR3 IGF-I/IGF-1 is a tag-free human IGF-1 analog (83 amino acids, E51R) with a 13-amino-acid N-terminal LR3 extension. Expressed in E. coli and provided as a high-purity preparation, it is intended for in vitro applications such as cell culture supplementation, signaling pathway studies, and biochemical assays requiring an IGF-1 analog with prolonged receptor activity.

  • Recombinant human IGF-1 analog with LR3 N-terminal extension and E51R substitution.
  • Tag-free, E. coli expressed protein.
  • High purity suitable for biochemical and cell culture applications.
  • Supplied in small research pack sizes including 10 μg.
  • Approximate length 83 amino acids and molecular weight ~9-11 kDa.
  • Suitable for signaling and proliferation assays in vitro.

Catalog No. 50-001-84132 Supplier Medchemexpress LLC Supplier No. HYP7078310UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant LR3 IGF-I/IGF-1 is a tag-free human IGF-1 analog (83 amino acids, E51R) with a 13-amino-acid N-terminal LR3 extension. Expressed in E. coli and provided as a high-purity preparation, it is intended for in vitro applications such as cell culture supplementation, signaling pathway studies, and biochemical assays requiring an IGF-1 analog with prolonged receptor activity.

  • Recombinant human IGF-1 analog with LR3 N-terminal extension and E51R substitution.
  • Tag-free, E. coli expressed protein.
  • High purity suitable for biochemical and cell culture applications.
  • Supplied in small research pack sizes including 10 μg.
  • Approximate length 83 amino acids and molecular weight ~9-11 kDa.
  • Suitable for signaling and proliferation assays in vitro.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.