Learn More
Yamato Scientific America, Inc. Yamato Digital Laboratory Stirrer, Max speed of 1000 rpm. Complete with stand and rod, stirring shaft and 4-blade propeller. 100V-125V
Catalog No. 501898199 Shop All Yamato Scientific America Inc. Products
Encompass_Preferred
Digital laboratory stirrer with speed range of 100~1000 rpm for medium viscosity samples. DC brushless motor considered superior in safety as there are no brushes to cause sparks and no brush replacement required. Direct-drive system reduces noise and require low maintenance. Achieves high torque enabling stirring of high viscosity solution. Digital tachometer for easy speed setting and confirmation. Load on the stirring shaft can be monitored by LED2 display. An overload lamp turns on when exceeding the maximum load, stopping the motor automatically. Revolution feedback control function can maintain the setting rate despite change of load. Set includes stand, rod, stainless steel shaft and 75mm 4-blade propeller.
Catalog No. 50-189-8199 Supplier Yamato Scientific America, Inc. Supplier No. LR500BA
Digital laboratory stirrer with speed range of 100~1000 rpm for medium viscosity samples. DC brushless motor considered superior in safety as there are no brushes to cause sparks and no brush replacement required. Direct-drive system reduces noise and require low maintenance. Achieves high torque enabling stirring of high viscosity solution. Digital tachometer for easy speed setting and confirmation. Load on the stirring shaft can be monitored by LED2 display. An overload lamp turns on when exceeding the maximum load, stopping the motor automatically. Revolution feedback control function can maintain the setting rate despite change of load. Set includes stand, rod, stainless steel shaft and 75mm 4-blade propeller.