Learn More
Aviva Systems Biology Corp LRAT Antibody - N-terminal region (ARP59785_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59785_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LRAT. It was validated on Western Blot. . Alternate names: MGC33103, LCA14ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-823