Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc 10mL, 1/16in Sieve Plate 20m

Catalog No. 502397970 Shop All Phenomenex Products
Encompass_Preferred

LRC Filtration Column 10mL FCL010

Catalog No. 50-239-7970 Supplier Phenomenex Inc Supplier No. FCL010
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LRC Filtration Column 10mL FCL010

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.