Learn More
Medchemexpress LLC HY-100524 5mg Medchemexpress, LRE1 CAS:1252362-53-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1005245MG
Encompass_Preferred
Medchemexpress, HY-100524 5mg LRE1 CAS:1252362-53-0 LRE1 is a specific and allosteric inhibitor of soluble adenylyl cyclase. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9461